missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRISP3 Monoclonal Antibody (295220)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561150
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
Cysteine-rich secretory protein 3 (CRISP-3; also known as SGP28) is a glycoprotein that belongs to the family of cysteine-rich secretory proteins (CRISPs) which was originally discovered in human neutrophilic granulocytes. CRISP-3 is also widely distibuted in exocrine glands (salivary glands, pancreas and prostate), eosinophilic granulocytes and to a lower level in epididymis, ovary, thymus and colon. The presence of CRISP-3 in neutrophils, eosinophils and in exocrine secretions indicates a role in innate host defense. The antibody has been raised against recombinant C-terminally truncated form of CRISP-3 and recognizes both the N-glycosylated and non-glycosylated form of the mature protein.
Spezifikation
| CRISP3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P54108 | |
| Crisp3 | |
| Mouse myeloma cell line NS0-derived recombinant human CRISP-3, Asn21-Tyr245 (Ser106Pro & Ala134Ser). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 295220 | |
| Unconjugated | |
| Crisp3 | |
| 32 kDa epididymal protein; Acidic epididymal glycoprotein; Acidic epididymal glycoprotein 2; acidic epididymal glycoprotein D/E; Aeg; Aeg2; Aeg-2; Crisp1; Crisp-1; Crisp3; CRISP-3; CRISP-3MGC126588; CRS3; cysteine rich secretory protein 3; cysteine-rich secretory protein 1; Cysteine-rich secretory protein 3; cysteine-rich secretory protein-3; dJ442L6.3; epididymal glycoprotein; protein D; protein E; protein IV; SCP; SCP 2; SGP28; Sialoprotein; specific granule protein (28 kDa); specific granule protein of 28 kDa; Sperm-coating glycoprotein; sperm-coating glycoprotein 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur