Learn More
SPEX Certiprep™ Cryogenic Mill
Marke:  SPEX Certiprep™ 6870-230
Artikelnummer. 12839045
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Fully programmable freezer mill that can store up to 10 user-defined methods for simple, fast and error free recall. A full colour, detachable control screen is intuitive to use for quick and easy set-up and operation. A truly cryogenic mill that cools samples in liquid nitrogen and then pulverises them by magnetically shuttling a steel impactor between two steel end plugs within a sealed vial. Suitable for samples up to 100g. This mill is supplied with a choice of one included accessory pack: if using large vials only, specify pack 6870L; if running small vials specify 6870S; if using midsize vials select, 6870M. If running more than one size of vial, more than one of these accessory packs will be needed, so order them as required.
  • Direct immersion in liquid nitrogen for optimum performance
  • Insulated case, nitrogen sensor and safety interlock ensures safe operation
  • Intuitive touch screen for setting grinding cycles, time, impact rate and cooling times
  • Sealed vials are suitable for volatile, temperature-sensitive and hazardous samples, and help eliminate cross-contamination
  • Pre-cooling chamber allows sample chilling while active chambers are running, helping maximise productivity
  • Magnet-driven impactor is only moving part, so unit is practically maintenance-free
  • Chamber takes large grinding vials for samples up to 100g, or four small grinding vials (6807 multivial adaptor needed)
  • Additionally, the chamber can accommodate four microvial sets, meaning that up to 12 samples can be ground simultaneously
  • Grinding vials are available in a variety of materials, including polycarbonate, stainless steel and Cr-free, to suit a range of applications
  • Supplied with 6804 extractor/vial opener and large vial rack
  • Optional 6820 liquid nitrogen autofill is available
Applications include: DNA/RNA extractions, forensics, materials testing, pharmaceutical analysis, drug testing, RoHS/WEEE and volatiles extraction. Typical samples include all types of plant and animal tissues, plastics and polymers, yeast, pharmaceuticals, food products, electronic components, textiles, hair, teeth and bones.
Dimensions (w x d x h), mm: 520 x 550 x 460Net weight: 24kg Gross weight: 32kgTimer/controls: Stores ten user-defined grinding programmesCertifications: CE approvedVoltage: 230V, 50Hz