Learn More
Gonotec™ Gefrierpunktosmometer 3000
Artikelnummer. 15104726
Change view
Click to view available options
Displaytyp:
LCD-Touchscreen
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Displaytyp unitSize
15104726 LCD-Touchscreen 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15104726 Lieferant Gonotec™ Lieferanten-Nr. 32
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen
The Gonotec™ Osmomat 3000 Single Sample Freezing Point Osmometer is especially designed for routine measurements in the medical field and is also very suitable for measurements in research and industry.
  • Determines the total osmolality of aqueous solutions
  • Requires very small sample volumes and can thus be applied for extreme measuring tasks
  • Rapidity allows serial measurements in a very short time
  • Can be controlled easily and comfortably via a touch-screen display
  • Step-by-step user guidance through all measurement functions
  • QM assistance for the laboratory supervisor
  • 2- or 3-point calibration
  • Results are sent to the optional built-in printer in document-ready format
  • For data transfer to a PC it can be connected via USB or RS232
  • Last result remains available for reading even after automatic switching to stand-by mode
  • Robust design of the measurement equipment makes this unit easy to handle and maintain
  • Choice of language menu
  • Automatic calibration by using Gonotec calibration standards
Spezifikation
Displaytyp LCD-Touchscreen
Kühlmittel By means of two separate peltier cooling systems with heat dissipation by air; crystallization process by means of the tip of a stainless steel needle covered with ice crystals which is automatically controlled
Zur Verwendung mit (Anwendung) General medicine, routine and research, forensic medicine, electron microscopy, physiology, clinical laboratories, paediatrics, urology and more
Messzeit 60 sec.
Bereich 0 to 3000 mOsmol/kg H2O
Wiederholgenauigkeit <±2 digit (0...400) mOsmol/kg H2O
< ±0.5% (400...1500) mOsmol/kg H2O
< ±1.0% (1500...3000) mOsmol/kg H2O
Auflösung 1 mOsmol/kg H2O
Probengröße 50 μl (optional 15 μl)
Betriebstemperatur (metrisch) 10°C to 35°C
Gewicht (metrisch) 6.4 kg
Elektrische Anforderungen 100/240 V 50/60 Hz 45 VA