missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CS Citrate Synthase Antibody (CL2553) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-36773MFV500
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CS Citrate Synthase Monoclonal antibody specifically detects CS Citrate Synthase in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Spezifikation
| CS Citrate Synthase | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| citrate synthase, citrate synthase, mitochondrial, EC 2.3.3, EC 2.3.3.1 | |
| This antibody was developed using a recombinant protein derived from O75390, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 1431 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL2553 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction