missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTGF/CCN2 Antibody (CL5339) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-61415G
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CTGF/CCN2 Monoclonal antibody specifically detects CTGF/CCN2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
| CTGF/CCN2 | |
| Monoclonal | |
| DyLight 488 | |
| CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
| This antibody was developed using a recombinant protein derived from P29279, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity | |
| 1490 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL5339 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur