missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTH Monoclonal Antibody (24GB5620)
Description
CTH Monoclonal Antibody for Western Blot, Flow
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
Specifications
| Antigen | CTH |
| Anwendungen | Flow Cytometry, Western Blot |
| Klassifikation | Monoclonal |
| Klon | 24GB5620 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | CTH |
| Gen-Zugriffsnummer | P18757, P32929 |
| Gen-Alias | 0610010I13Rik; AI314617; BC019483; CSE; Cth; cth protein; CTL target antigen; Cys3; cystathionase; cystathionase (cystathionine gamma-lyase); cystathionase isoform 1; cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; cysteine-protein sulfhydrase; EGK_00877; fb58d08; gamma-cystathionase; H; homoserine deaminase; homoserine dehydratase; LOC102117863; MGC9471; PRB-RA; probasin-related antigen; QccE-22305; synthase; Unknown (protein for MGC:85785); wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785 |
| Gensymbole | CTH |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?