Learn More
Ibidi Culture-Insert 2 Well 24 ibiTreat
Beschreibung
- 24 x 2 well silicone insert with a defined cell-free gap of 500 μm
- suitable for wound healing, migration assays, 2D invasion assays, and co-cultivation of cells
- in a μ-Plate 24 Well with #1.5 Polymer Coverslip bottom
- tissue culture treated
- sterilized
Spezifikation
Spezifikation
| Typ | Kultureinsatz |
| Menge | 3 |
| Wells | 2 |
| Sterilität | Steril |
| Anzahl Wells | 2 |