Learn More
Invitrogen™ Custom Primer, Plate
Product Code. 10500163
Change view
Click to view available options
Quantity:
200 nmol
25 nmol
50 nmol
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10500163 25 nmol Each
10154082 50 nmol Each
10367112 200 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10500163 Supplier Invitrogen™ Supplier No. 10629152
Please to purchase this item. Need a web account? Register with us today!
Custom Primer, Plate
Order multiple custom DNA oligos in plate format by selecting either 96 or 384-wells formats. Simply download either the 96-well Excel template or 384-well Excel template, enter your sequence and plate location information, and upload in the appropriate format.
TRUSTED_SUSTAINABILITY
Specifications
Description Primer, 25 nmol, Order Multiple Custom DNA Oligos in Plate Format by Selecting Either 96 or 384-wells Formats, Simply Download Either the 96-well Excel Template or 384-Well Excel Template, Enter Your Sequence and Plate Location Information and Upload in the Appropriate Format
Product Type Primer
Content And Storage Room Temperature
Quantity 25 nmol