Learn More
Invitrogen™ CXC-ELR Biosimilar Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
500.00 € - 1262.00 €
Spezifikation
| Antigen | CXC-ELR Biosimilar |
|---|---|
| Konzentration | 1 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30282432
![]() |
Invitrogen™
MA558953 |
100 μg |
500.00 €
100 Mikrogramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
|
30282469
|
Invitrogen™
MA558954 |
1 mg |
1262.00 €
1 Milligramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Lilly patent anti-Pan-ELR+.
CXC chemokines are a subgroup of the chemokine superfamily distinguished by the presence of a C-X-C motif, where 'X' is any amino acid, and they play pivotal roles in immune responses by guiding the migration of leukocytes. A critical subset of these chemokines contains the glutamic acid-leucine-arginine (ELR) motif, which significantly influences angiogenesis and immune cell recruitment. These ELR+ CXC chemokines, including CXCL1, CXCL2, CXCL3, CXCL5, CXCL6, CXCL7, and CXCL8, primarily bind to CXCR1 and CXCR2 receptors. These receptor-ligand interactions activate intracellular signaling pathways essential for neutrophil chemotaxis, tumorigenesis, and wound healing. The structural basis of these interactions and their biological implications, including their roles in pathological conditions such as cancer and inflammatory diseases, have been extensively studied, highlighting their significance in therapeutic targeting.Spezifikation
| CXC-ELR Biosimilar | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG4SP | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.