missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexene-d10, CDN
Click to view available options
Menge:
0.5 g
1 g
Packungsgröße:
0.50 Gramm
1 Gramm
Spezifikation
Spezifikation
| Chemischer Name oder Material | Cyclohexene-d10 |
| CAS | 1603-55-0 |
| Summenformel | C6 D10 |
| Menge | 1 g |
| Synonym | Cyclohexene-1,2,3,3,4,4,5,5,6,6-d10; |
| InChI-Formel | InChI=1 S/C6H10/c1-2-4-6-5-3-1/h1-2 H,3-6H2/i1D,2 D,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C1=C([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC-Name | 1,2,3,3,4,4,5,5,6,6-decadeuteriocyclohexene |
| Molekulargewicht (g/mol) | 92.21 |
| Formelmasse | 92.141 g/mol |
| Mehr anzeigen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction