Learn More
R&D Systems™ Cynomolgus Monkey CD94 (CAE51857.1) VersaClone cDNA
Spezifikation
Spezifikation
| Gen | CD94 |
| Zugriffsnummer | CAE51857.1 |
| Gen-ID (Entrez) | 3824 |
| Gen-Alias | CD94, CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KLRD1, KP43, NK cell receptor |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Cynomolgus Monkey |
| Produktlinie | R&D Systems |
| Menge | 10 μg |