missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin S Monoclonal Antibody (201805)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA560965
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.
Spezifikation
| Cystatin S | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P01036 | |
| CST4 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin S, Ser21-Ala141. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 201805 | |
| Unconjugated | |
| CST4 | |
| CST4; cystatin 4; cystatin S; cystatin-4; Cystatin-S; cystatin-SA-III; Salivary acidic protein 1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1472 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur