Learn More
Cytochrome c Antibody (CYCS/3128R), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-08598H
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cytochrome c Monoclonal antibody specifically detects Cytochrome c in Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spezifikation
| Cytochrome c | |
| Monoclonal | |
| HRP | |
| CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
| Recombinant full-length human Cytochrome c protein (Uniprot: P99999) | |
| 100 μg | |
| Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
| 54205 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CYCS/3128R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Amphibian, Canine | |
| Purified |