missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytochrome C Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA581161
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Cytochrome c.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
Spezifikation
| Cytochrome C | |
| Polyclonal | |
| Unconjugated | |
| Cycs | |
| CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 54205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| P99999 | |
| Cycs | |
| A synthetic peptide corresponding to the C-terminus of the Human Cytochrome c. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur