Learn More
Invitrogen™ Cytochrome C1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA587907
Beschreibung
Immunogen sequence: SDLELHPPSY PWSHRGLLSS LDHTSIRRGF QVYKQVCASC HSMDFVAYRH LVGVCYTEDE AKELAAEVEV QDGPNEDGEM FMRPGKLFDY FPKPYPNSEA ARAANNGALP PDLSYIVRAR HGGEDYVFSL LTGYCEPPTG VSLREGLYFN PYFPGQAIAM APPIYTDVLE FDDGTPATMS QIAKDVCTFL RWASEPEHDH RKRMGLK; Positive Samples: A-549, A375, LO2, 22Rv1, Mouse brain, Mouse kidney, Mouse liver, Mouse heart, Rat liver; Cellular Location: Intermembrane side, Mitochondrion inner membrane, Single-pass membrane protein.
This is the heme-containing component of the cytochrome b-c1 complex, which accepts electrons from Rieske protein and transfers electrons to cytochrome c in the mitochondrial respiratory chain.
Spezifikation
| Cytochrome C1 | |
| Polyclonal | |
| Unconjugated | |
| CYC1 | |
| 2610002H19Rik; AA408921; Complex III subunit 4; complex III subunit IV; Cyc1; Cytochrome b-c1 complex subunit 4; cytochrome c1; cytochrome c-1; cytochrome c1, heme protein, mitochondrial; MC3DN6; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; UQCR4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1537, 300047, 66445 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08574, Q9D0M3 | |
| CYC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 85-291 of human CYC1 (NP_0019072). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.