missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 14 Antibody (CL14565) [Alexa Fluor™ 647], Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Spezifikation
Spezifikation
| Antigen | Cytokeratin 14 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | CL14565 |
| Konjugat | Alexa Fluor 647 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P02533, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?