Learn More
Cytokeratin 14 Antibody (CL14568) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP3-26869PE
Artikelnummer. 30298346
  • 770.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Cytokeratin 14
Monoclonal
PE
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ
This antibody was developed using a synthetic peptide derived from P02533, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers
3861
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
CL14568
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen