missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Antibody (KRT19/800), Novus Biologicals™

Mouse Monoclonal Antibody

299.00 € - 509.00 €

Spezifikation

Antigen Cytokeratin 19
Klon KRT19/800
Konzentration 0.2mg/mL
Verdünnung Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Anwendungen Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
18003386
Unterlagen ansehen
Novus Biologicals
NBP2-44827-0.02MG
0.02 mg
299.00 €
0.02 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
18711554
Unterlagen ansehen
Novus Biologicals
NBP2-44827-0.1mg
509.00 €
0.10 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Spezifikation

Spezifikation

Cytokeratin 19
0.2mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Mouse
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366
KRT19
IgG1 κ
Protein A or G purified
Recognizes a protein of 40kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.
KRT19/800
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Monoclonal
Purified
RUO
Human, Mouse, Rat
P08727
3880
Recombinant human KRT19 protein
Primary
Store at 4C.
40 kDa
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis