missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 20 Antibody (E19-I), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP2-21840-200UL
This item is not returnable.
View return policy
Description
Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 20 | |
| Monoclonal | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
| CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| E19-l | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200 | |
| P35900 | |
| KRT20 | |
| Peptide derived from C-terminal region of human cytokeratin 20. | |
| 200 μL | |
| Cytoskeleton Markers | |
| 54474 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction