missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, Low Molecular Weight, Ab-1 Mouse Monoclonal Antibody, Epredia™
Tienda Epredia ProductosDescripción
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. The basic keratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. Members of acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with cell type, differentiation status and environment. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Host Species: Mouse
Clone: AE1
Isotype: IgG1
Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Epitope: Not determined
Immunogen: Human epidermal Cytokeratin
Positive Control: Skin, Squamous cell CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Especificaciones
Especificaciones
| Antigen | Cytokeratin, Low Molecular Weight Ab-1 |
| Anwendungen | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | AE1 |
| Konzentration | 200μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Immunogen | Human epidermal keratin |
| Menge | 100 μL |
| Regulatorischer Status | IVD |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?