Learn More
Thermo Scientific Chemicals Cytosin, ≥ 98 %
Beschreibung
Cytosine is a nucleoside used for proteomics research. It is also used as an enzyme substrate or precursor of effector molecules such as cytosine sugars.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cytosin ist ein Nukleosid, das für die Proteomikforschung verwendet wird. Es wird auch als Enzymsubstrat oder Vorläufer von Effektormolekülen wie Cytosinzucker verwendet.
Löslichkeit
Klare bis sehr leicht dunstige, farblose bis schwach gelbe Lösung bei 50 mg/ml in 0.5 M HCL.
Hinweise
Stabil unter empfohlenen Lagerbedingungen. Nicht kompatibel mit Oxidationsmitteln.
Specifications
Specifications
| Chemischer Name oder Material | Cytosine |
| Schmelzpunkt | >300°C |
| Menge | 5 g |
| Beilstein | 2637 |
| Merck Index | 14,2795 |
| Löslichkeitsinformationen | Clear to very slightly hazy colorless to faint yellow solution at 50mg/ml in 0.5 M HCL. |
| IUPAC-Name | 6-Amino-1H-Pyrimidin-2-on |
| Formelmasse | 111.1 |
| Reinheit (%) | ≥98% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.