missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (DD2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB110-8376UV
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA, Immunoassay, Sandwich ELISA
Spezifikation
| D-Dimer | |
| Monoclonal | |
| DyLight 350 | |
| D Dimer | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer. | |
| 0.2 mL | |
| Extracellular Matrix | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoassay, Sandwich ELISA | |
| DD2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Spot an opportunity for improvement?Share a Content Correction