missing translation for 'onlineSavingsMsg'
Learn More

D,L-Sulforaphan, TRC

Artikelnummer. 17938304
Ansicht ändern
Click to view available options
Menge:
25 mg
250 mg
1 g
Unit Size:
1 Gramm
25 Milligramm
250 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
17938304 25 mg 25 Milligramm
17959724 250 mg 250 Milligramm
17969344 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938304 Supplier Toronto Research Chemicals Supplier No. TRCS69911525MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4478-93-7
Summenformel C6 H11 N O S2
Molekulargewicht (g/mol) 177.29
Synonym Isothiocyanic acid, 4-(methylsulfinyl)butyl ester (8CI), Sulforaphan (6CI), 1-Isothiocyanato-4-(methylsulfinyl)butane, 4-Methylsulfinylbutyl isothiocyanate, BroccoPhane, Detoxophane, Sulforaphane, Butane, 1-isothiocyanato-4-(methylsulfinyl)-
IUPAC-Name 1-Isothiocyanato-4-Methylsulfinylbutan
SMILES CS(=O)CCCCN=C=S

Specifications

Chemischer Name oder Material DL-Sulforaphane
Menge 25 mg
InChI-Formel InChI=1S/C6H11NOS2/c1-10(8)5-3-2-4-7-6-9/h2-5H2,1H3
Formelmasse 177.0282
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.