missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGNR/CD299/CLEC4M Antibody (MM0241-2U25), PE, Novus Biologicals™
Alle ansehen Bio Techne ProdukteDescription
DC-SIGNR/CD299/CLEC4M Monoclonal specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | DC-SIGNR/CD299/CLEC4M |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | MM0241-2U25 |
| Konjugat | PE |
| Verdünnung | Flow Cytometry |
| Gen-Zugriffsnummer | NP_001138382 |
| Gen-Alias | CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR |
| Gensymbole | CLEC4M |
| Wirtsspezies | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?