missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDRGK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21445-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
21445-1-AP is a rabbit polyclonal antibody raised against the full-length of human DDRGK1.
Protein which interacts with the E3 UFM1-protein ligase UFL1 and one of its substrates TRIP4 and is required for TRIP4 ufmylation. Through TRIP4 ufmylation may regulate nuclear receptors-mediated transcription. May play a role in NF-kappa-B-mediated transcription through regulation of the phosphorylation and the degradation of NFKBIA, the inhibitor of NF-kappa-B. May also play a role in the cellular response to endoplasmic reticulum stress.Spezifikation
| DDRGK1 | |
| Polyclonal | |
| Unconjugated | |
| DDRGK1 | |
| C20orf116, DDRGK domain containing 1, DDRGK1, UFBP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 65992, 77006 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80WW9, Q96HY6 | |
| DDRGK1 | |
| DDRGK1 Fusion Protein Ag13742 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur