missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Defensin alpha 5 Antibody (8C8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
231.00 € - 518.00 €
Spezifikation
| Antigen | Defensin alpha 5 |
|---|---|
| Klon | 8C8 |
| Konzentration | 2.35 mg/mL |
| Verdünnung | Western Blot 1:1000, Flow Cytometry 1 ug per million cells, ELISA reported in scientific literature, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 30612153), Dot Blot reported in scientific literature |
| Anwendungen | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18226760
|
Novus Biologicals
NB110-60002SS |
0.025 mL |
231.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18760754
|
Novus Biologicals
NB110-60002 |
518.00 €
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | ||||||
Beschreibung
Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Spezifikation
| Defensin alpha 5 | |
| 2.35 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| Q01523 | |
| 1670 | |
| Full-length human Defensin alpha 5 [Swiss-Prot# Q01523]. The propeptide is the full-length protein. In the mature form, it is cleaved at residue 63 (alanine). | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 10 kDa |
| 8C8 | |
| Western Blot 1:1000, Flow Cytometry 1 ug per million cells, ELISA reported in scientific literature, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 30612153), Dot Blot reported in scientific literature | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DEF5defensin, alpha 5, preproprotein, defensin 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728 | |
| DEFA5 | |
| IgG2b | |
| Protein G purified | |
| This is specific for HD5. It does not cross-react with HD6, lysozyme or sPLA2. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts