missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ delta Catenin Monoclonal Antibody (24GB5775)

Beschreibung
delta Catenin Monoclonal Antibody for Western Blot
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
Specifications
Specifications
| Antigen | delta Catenin |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | 24GB5775 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | CTNND1 |
| Gen-Zugriffsnummer | O60716, P30999 |
| Gen-Alias | AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN |
| Gensymbole | CTNND1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?