Learn More
Chemsplash Delta Coverall (Sterile Irradiated)

Descripción
Chemsplash Delta 67 Coverall
- 67GSM Microporous Laminate Fabric
- Sterile Irradiated, Double Bagged and Detex Sticker
- Low Linting Knitted Cuffs with Thumb Loops
- Elasticated Three Piece Hood
- Two Way Zip
- Adhesive Zip Flap
- Elasticated Back and Ankles
- Reinforced Bound Seam at Crotch Area
- Non Linting and Anti-Static
- Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Delta 67 Coverall is made from soft, comfortable 67gsm microporous laminate fabric, meeting Cat III Type 5B/6B. It is Sterile Irradiated, double-bagged, and comes with a Detex sticker for identification. The coverall is compliant with EN1149-5:2008 (Antistatic) and EN1073-2 (Nuclear Particle). Chemsplash Delta is ideal for prolonged wear, offering improved breathability while maintaining an effective hazard barrier. This coverall is suitable for a wide range of applications, with an elasticated back, hood, cuffs, ankles, and reinforced seams for added durability. It conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9.
Especificaciones
Especificaciones
| Produkttyp | Cleanroom Coveralls |
| Größe | Small |
| Geschlecht | Unisex |
| Material | Microporous Fabric |
| Farbe | White |
| Einwegprodukt | Single-use |
| Taillenform | Elastic |
| Knöchelform | Elastic |
| Stulpenform | Knit, Thumb Loop |
| Flaschenhalstyp | Hooded |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.