missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desethylatrazine-d7 (iso-propyl-d7), CDN
Click to view available options
Menge:
0.01 g
0.05 g
Packungsgröße:
0.01 Gramm
0.05 Gramm
Spezifikation
Spezifikation
| Chemischer Name oder Material | 2-Amino-4-(isopropyl-d7-amino)-6-chloro-triazine |
| CAS | 1216649-31-8 |
| Summenformel | C6 D7 H3 Cl N5 |
| Menge | 0.05 g |
| Synonym | Atrazine-desethyl D7 (Isopropyl D7), 6-Chloro-N2-[1-(methyl-d3)ethyl-1,2,2,2-d4]-1,3,5-triazine-2,4-diamine, 6-Chloro-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine, D7 deethylatrazine |
| InChI-Formel | InChI=1 S/C6H10ClN5/c1-3(2)9-6-11-4(7)10-5(8)12-6/h3H,1-2H3,(H3,8,9,10,11,12)/i1D3,2D3,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Nc1nc(N)nc(Cl)n1)C([2 H])([2 H])[2 H] |
| IUPAC-Name | 6-chloro-2-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine |
| Molekulargewicht (g/mol) | 194.6732 |
| Formelmasse | 194.1064 g/mol |
| Mehr anzeigen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction