missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHCR24 Recombinant Rabbit Monoclonal Antibody (HL3253)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556870
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DHCR24 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
DHCR24 is a flavin adenine dinucleotide (FAD)-dependent oxidoreductase which catalyzes the reduction of the delta-24 double bond of sterol intermediates during cholesterol biosynthesis. This protein contains a leader sequence that directs it to the endoplasmic reticulum membrane. Missense mutations in this gene have been associated with desmosterolosis. Also, reduced expression of its gene occurs in the temporal cortex of Alzheimer disease patients and overexpression has been observed in adrenal gland cancer cells.
Spezifikation
| DHCR24 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q15392, Q5BQE6, Q8VCH6 | |
| Dhcr24 | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human Seladin 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL3253 | |
| Unconjugated | |
| Dhcr24 | |
| 2310076D10Rik; 24-dehydrocholesterol reductase; 3 beta-hydroxysterol delta 24-reductase; 3-beta-hydroxysterol delta-24 reductase; 3-beta-hydroxysterol delta-24-reductase; 5830417J06Rik; DCE; Delta(24)-sterol reductase; desmosterol-to-cholesterol enzyme; DHCR24; Diminuto/dwarf1 homolog; KIAA0018; mKIAA0018; Nbla03646; seladin 1; SELADIN1; seladin-1; selective AD indicator 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1718, 298298, 74754 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur