missing translation for 'onlineSavingsMsg'
Learn More

Dienogest Antibody (C3), Novus Biologicals™

Product Code. 30281061 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
100 μL
Unit Size:
100 Mikroliter
Product Code. Menge unitSize
30281061 100 μL 100 Mikroliter
1 options
This item is not returnable. View return policy

Product Code. 30281061

Brand: Novus Biologicals NBP343735

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dienogest Monoclonal antibody specifically detects Dienogest in Nonspecies specific samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dienogest
Anwendungen ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
Klassifikation Monoclonal
Klon C3
Konzentration 1 mg/mL
Konjugat Unconjugated
Verdünnung ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunohistochemistry-Frozen 5-20 μg/mL
Zusammensetzung 0.01 M PBS, pH 7.4, 50% glycerol
Wirtsspezies Mouse
Immunogen BSA - Dienogest
Reinigungsverfahren Protein A or G purified
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Non-species specific
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.