missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Antibody, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Polyclonal antibody specifically detects Dimethylarginine Dimethylaminohydrolase 1/DDAH1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spezifikation
Specifications
| Antigen | Dimethylarginine Dimethylaminohydrolase 1/DDAH1 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry-Paraffin 1:100-1:400 |
| Zusammensetzung | PBS (pH 7.0) |
| Gen-Alias | DDAH-1, DDAHdimethylargininase-1, DDAHI, Dimethylargininase-1, dimethylarginine dimethylaminohydrolase 1FLJ25539, EC 3.5.3.18, FLJ21264, N(G), N(G)-dimethylarginine dimethylaminohydrolase 1, NG, NG-dimethylarginine dimethylaminohydrolase |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human Dimethylarginine Dimethylaminohydrolase 1/DDAH1 fragment. |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?