missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DKKL1 Monoclonal Antibody (192425)

Rat Monoclonal Antibody
Marke: Invitrogen™ MA560994
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
The dickkopf protein family interacts with the Wnt signaling pathway and its members are characterized by two conserved cysteine-rich domains. This gene encodes a secreted protein that has high similarity to the N-terminus of the dickkopf-3 protein and moderate similarity to that protein's C-terminus though the C-terminal cysteine residues are not conserved.
Spezifikation
| DKKL1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9QZL9 | |
| DKKL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Soggy-1/DkkL1, Leu21-Leu230. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 192425 | |
| Unconjugated | |
| DKKL1 | |
| AI414495; Cancer/testis antigen 34; CT34; dickkopf like acrosomal protein 1; dickkopf-like 1; dickkopf-like 1 (soggy); dickkopf-like protein 1; Dkkl1; mSgy; Protein soggy-1; RGD1563523; SGY; SGY1; SGY-1; Soggy; soggy 1; soggy-1; testicular secretory protein Li 15; UNQ735/PRO1429 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 50722 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur