missing translation for 'onlineSavingsMsg'
Learn More

DNA Ligase III Antibody (1F3), Novus Biologicals™

Artikelnummer. 18034804 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μL
Unit Size:
100 Mikroliter
Product Code. Menge unitSize
18034804 100 μL 100 Mikroliter
1 options
This item is not returnable. View return policy

Product Code. 18034804

Brand: Novus Biologicals NBP141190

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNA Ligase III Monoclonal specifically detects DNA Ligase III in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase III
Anwendungen Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Blocking Assay, Proximity Ligation Assay
Klassifikation Monoclonal
Klon 1F3
Konzentration 1.0mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay, Block/Neutralize
Gen-Alias DNA ligase 3, DNA ligase III, EC 6.5.1.1, LIG2, ligase II, DNA, ATP-dependent, ligase III, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 3
Gensymbole LIG3
Wirtsspezies Mouse
Immunogen Recombinant full-length human DNA Ligase III-b.
Molekulargewicht des Antigens 100 kDa
Reinigungsverfahren Immunogen affinity purified
Menge 100 μL
Regulatorischer Status RUO
Forschungsgebiet Base Excision Repair, Cancer, DNA Repair
Primär oder sekundär Primary
Gen-ID (Entrez) 3980
Testspezifität This antibody will recognize human DNA ligase III.
Zielspezies Human, Mouse, Chicken
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.