missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Ligase III Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15858463
Change view
Click to view available options
Menge:
100 μL
Unit Size:
100 Mikroliter
Product Code. Menge unitSize
15858463 100 μL 100 Mikroliter
1 options
This item is not returnable. View return policy

Product Code. 15858463

Brand: Invitrogen™ PA570055

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100 kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97 kDa (beta form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase III
Anwendungen Western Blot
Klassifikation Polyclonal
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 2% sucrose and 0.09% sodium azide
Gen LIG3
Gen-Zugriffsnummer 0
Gen-Alias D11Wsu78e; DNA ligase (ATP) 3; DNA ligase 3; DNA ligase III; hypothetical protein; LIG2; LIG3; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 3; Unknown (protein for MGC:134139)
Gensymbole LIG3
Wirtsspezies Rabbit
Immunogen synthetic peptide corresponding to a region of rat DNA Ligase III.
Reinigungsverfahren Affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 303369
Zielspezies Rat
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.