Learn More
DNA Polymerase epsilon subunit 3 Antibody - BSA Free, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
DNA Polymerase epsilon subunit 3 Polyclonal antibody specifically detects DNA Polymerase epsilon subunit 3 in Human, Mouse samples. It is validated for Western Blot, Gene Knock-Out
Spezifikation
Spezifikation
| Antigen | DNA Polymerase epsilon subunit 3 |
| Anwendungen | Western Blot, Gene Knock-Out |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500 - 1:2000, Knockout Validated |
| Zusammensetzung | PBS with 50% glycerol, pH7.3. |
| Gen-Alias | Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human DNA Polymerase epsilon subunit 3 (NP_059139.3). MAERPEDLNLPNAVITRIIKEALPDGVNISKEARSAISRAASVFVLYATSCANNFAMKGKRKTLNASDVLSAMEEMEFQRFVTPLKEALEAYRREQKGKKEASEQKKKDKDKKTDSEEQDKSRDEDNDEDEERLEEEEQNEEEEVDN |
| Reinigungsverfahren | Affinity purified |
| Mehr anzeigen |