missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOC2A Polyclonal Antibody

Sheep Polyclonal Antibody
Marke: Invitrogen™ PA5149422
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2A is mainly expressed in brain and is suggested to be involved in Ca(2+)-dependent neurotransmitter release.
Spezifikation
| DOC2A | |
| Polyclonal | |
| Unconjugated | |
| DOC2A | |
| Doc2; DOC2A; Doc2-alpha; double C2 domain alpha; double C2, alpha; double C2-like domain-containing protein alpha; double C2-like domains, alpha | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 13446, 8448 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q14183, Q7TNF0 | |
| DOC2A | |
| E. coli-derived recombinant human Doc2 alpha , Met1-Lys114 (Gly48Ser). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur