missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
Spécification
| Antigen | DOG1/TMEM16A |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | rDG1/447 |
| Konjugat | HRP |
| Zusammensetzung | PBS |
| Gen-Alias | ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant full-length human DOG1/TMEM16A protein |
| Reinigungsverfahren | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?