missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody has been used in 5 publications
Marke: Novus Biologicals NBP2-60657
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Spezifikation
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| 0.05M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/ml BSA- Immunoglobulin and Protease free with 0.05% Sodium Azide | |
| Goat IgG whole molecule | |
| 1 mg | |
| Secondary | |
| Goat | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay | |
| Donkey | |
| Affinity Purified | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Chicken, Guinea Pig, Hamster, Horse, Mouse, Rabbit, and Rat Serum Proteins. No reaction was observed against Chicken, Guinea Pig, Hamster, Horse, Mouse, Rabbit or Rat Serum Proteins. Specificity was confirmed using ELISA minimal cross reactivity against the targets stated above. | |
| Store at 4C. Do not freeze. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur