Learn More
DiscoverX Dopamine
Artikelnummer. 17312489
Encompass
Change view
Click to view available options
Menge:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
17312489 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17312489 Lieferant DiscoverX Lieferanten-Nr. 921161
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Dopamine is an agonist for DRD1-DRD5. Provided as 10mg of dry powder.
Dopamine hydrochloride is an endogenous neurotransmitter with sympathomimetic properties that is a potent agonist of the dopamine receptors DRD1-5. Increases heartbeat and blood flow to the body's tissues, also playing a role in reward-motivated behavior. Addictive drugs tend to increase dopamine activity, and multiple nervous system diseases are associated with its dysfunction, including Parkinson's and schizophrenia. Precursor in noradrenalin and adrenalin biosynthesis. Provided as 10mg of dry powder.
Spezifikation
Menge 10 mg