Learn More
Invitrogen™ Dopamine Transporter Recombinant Rabbit Monoclonal Antibody (24GB2160)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557262
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Dopamine Transporter Recombinant Monoclonal Antibody for Western Blot, Flow
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Spezifikation
| Dopamine Transporter | |
| Recombinant Monoclonal | |
| 0.40 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q01959 | |
| Slc6a3 | |
| A synthesized peptide derived from human Dopamine Transporter (aa 150-215). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2160 | |
| Unconjugated | |
| Slc6a3 | |
| DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6531 | |
| -20°C | |
| Liquid |