Learn More
Socorex Dosys all glass 155 syringe, 100ml, Luer Lock, metal
Marke:  Socorex 155.05100
Dosys all glass 155 syringe, 100ml, Luer Lock, metal
Artikelnummer. 12653275
  • ONLINE EXCLUSIVE PRICE
    58.35 €
    46.20 € / 1 Stück
    Sparen 12.15 €
    20% Rabatt