Learn More
Gilac™ Double-wall Container with Stainless Steel Wheel Housing
Click to view available options
Kapazität (metrisch):
310 l
500 l
Packungsgröße:
1 Stück
Beschreibung
- Ideal for transporting crushed ice, prepared food and marinated items.
- Wide range of uses.
- No gaps nor overlaps and no sharp corners.
- Double-walled offering excellent thermal insulation and excellent mechanical resistance.
- Container comprising a drainage outlet with drain plug.
- Rotational molded container with 160 mm, polypropylene wheels with synthetic rubber tires (2 fixed and 2 pivoting) and stainless steel wheel housing.
- Stainless steel clevis recommended in humid environments (no damage to the bearings even in the presence of water).
- Can be stored on 800 x 1200 pallets, stackable upside down.
- Integral high-flow drainage.
- 1/4 turn angle stop valve optional extra.
- Volume in litres is equivalent to the container filled to the brim.
Spezifikation
Spezifikation
| Typ | Storage Tote |
| Material | HDPE |
| Farbe | Weiß |
| Form | Rechteckig |
| Kapazität (angloamerikanisch) | 81.9 gal. |
| Kapazität (metrisch) | 310 l |