missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DRD1 Recombinant Rabbit Monoclonal Antibody (HL2680)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556682
This item is not returnable.
View return policy
Description
DRD1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
Specifications
| DRD1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P21728, Q61616 | |
| DRD1 | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human Dopamine Receptor D1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2680 | |
| Unconjugated | |
| DRD1 | |
| C030036C15Rik; D(1A) dopamine receptor; D1 receptor; D1a; D1A dopamine receptor; D1a receptor; DADR; Dopamine 1 receptor; Dopamine D1 receptor; dopamine receptor D1; dopamine receptor D1A; Dopamine-1A receptor; Drd1; Drd-1; Drd1a; G protein-coupled receptor DRD1; Gpcr15 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13488, 1812 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction