Learn More
DRP1 Antibody (4E11B11), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-80702V
Artikelnummer. 18448797
  • ONLINE EXCLUSIVE PRICE
    597.00 €
    563.85 € / 0.10 Milliliter
    Sparen 33.15 €
    5% Rabatt
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
DRP1 Monoclonal antibody specifically detects DRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
DRP1
Monoclonal
DyLight 405
50mM Sodium Borate
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
4E11B11
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1
Partial recombinant human DRP1 (between residues 100-250) expressed in E. coli. [Uniprot: O00429]
0.1 mL
Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction
10059
Store at 4C in the dark.
IgG1
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen