missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB110-55288SS
189.55 EUR gültig bis 2025-12-16
BESTER Aktionspreis! Benutzen Sie den Promotions-Code "24090" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
DRP1 Polyclonal specifically detects DRP1 in Human, Mouse, Rat, Fish, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| DRP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1 | |
| Rabbit | |
| 81 kDa | |
| 0.025 mL | |
| Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction | |
| 10059 | |
| Human, Mouse, Rat, Fish, Primate | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1 mg/mL | |
| Western Blot 1:500, Simple Western 1:50, Flow Cytometry 1:1000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin 2.5 ug/ml | |
| O00429 | |
| DNM1L | |
| A synthetic peptide made to an internal region within residues 500-600 of the human DRP1 protein. [Swiss-Prot# O00429] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, primate, rat, mouse. Predicted to react with bovine based on 100% sequence homology. Fish reactivity reported in scientific literature (PMID: 25008790). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction