missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Selectin/CD62E Antibody (RM0034-9J12) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NB110-85473-0.025MG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| E-Selectin/CD62E | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:400 | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
| Rat | |
| Protein G purified | |
| Primary | |
| This is specific for mouse E-Selectin | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Flow Cytometry | |
| RM0034-9J12 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| SELE | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with cytokine stimulated-mouse endothelial membrane protein. | |
| 0.025 mg | |
| 6401 | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto