Learn More
Invitrogen™ eBioscience™ Concanavalin A (Con A) Solution (500X)
Click to view available options
Menge:
100 μl
4 x 100 μL
Packungsgröße:
100 Mikroliter
4 Stück
Beschreibung
Con A is isolated from the Jack bean, Canavlia ensiformis. It is a lectin that binds alpha-D-glucose and alpha-D-mannose moieties found in various glycoproteins, glycolipids, and sugars and is a potent leukocyte mitogen. This reagent is intended for use in in vitro activation of human and mouse leukocytes.
Reported Application
Functional Assays
Spezifikation
Spezifikation
| Format | Vial |
| Menge | 4 x 100 μL |
| Quelle | Canavlia Ensiformis |
| Form | Solution |
| Protein-Subtyp | Lectins |
| Produktlinie | eBioscience |
For Research Use Only. Not for use in diagnostic procedures.