missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EBP1 Recombinant Rabbit Monoclonal Antibody (24GB580)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557635
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
EBP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.
Spezifikation
| EBP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Pa2g4 | |
| 38kDa; AA672939; Cell cycle protein p38-2G4 homolog; Ebp1; ErbB-3 binding protein 1; ErbB3-binding protein 1; ErbB3-binding protein Ebp1; hG4 1; hG4-1; IRES-specific cellular trans-acting factor 45 kDa; ITAF45; MGC81621; MGC94070; Mpp1; p38 2G4; p38-2G4; Pa2g4; Plfap; proliferation-associated 2G4; proliferation-associated 2G4, 38kD; proliferation-associated 2G4, 38kDa; proliferation-associated protein 1; Proliferation-associated protein 2G4; protein p38-2G4; si:dz150i12.2; wu:fb19b11; wu:ft56d05; zgc:86732 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18813, 288778, 5036 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB580 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P50580, Q6AYD3, Q9UQ80 | |
| Pa2g4 | |
| A synthesized peptide derived from human EBP1 / PA2G4 (aa 300-394). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur