Learn More
Tuttnauer ELV-D Vertical Autoclave
Artikelnummer. p-200007392 Alle ansehen Tuttnauer Produkte
Click to view available options
Kapazität (metrisch):
31 l
65 l
85 l
160 l
Eigenschaften:
Dreiphasig
Dreiphasig, BioHazard-Filter
Dreiphasig, Wasserreservoir
Einzelphase
Einzelphase, Wasserreservoir
Wasserreservoir
Packungsgröße:
1 Stück
Artikelnummer. 17801077
Marke: Tuttnauer 2840ELVWR230D
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
The ELV-D Vertical Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Designed for your laboratory applications. The 5075ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
  • Advanced Control System - Take Advantage of Tuttnauer's state-of-the-art control system with multi-color display
  • Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
  • Optional Internal Printing - Documents date, time, temperature and pressure
  • Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
  • High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
  • Diverse Load Types - The 5075ELV-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Spezifikation
Kapazität (angloamerikanisch) 6.8 gal.
Kapazität (metrisch) 31 l
Tiefe (metrisch) Kammer 40 cm
Durchmesser (angloamerikanisch) Kammer 11.2 in.
Tiefe (angloamerikanisch) Kammer 16 in.
Durchmesser (metrisch) Kammer 28 cm
Länge (angloamerikanisch) 21.2 in.
Länge (metrisch) 53 cm
Modell 2840ELVWR-D
Produkttyp Autoclave, Top-loading
Temperatur (angloamerikanisch) 221°F to 280°F
Temperatur (metrisch) 105°C to 137°C
Eigenschaften Wasserreservoir